2-Aminopropanamide hydrochloride

Supplier: Matrix Scientific

052748-500MG 052748-2.5G
101900-880EA 153.85 USD
101900-880 101900-882
2-Aminopropanamide hydrochloride
2-Aminopropanamide hydrochloride

Hazard: Irritant

Formula: C₃H₉ClN₂O
MW: 124.57 g/mol
MDL Number: MFCD00066144
CAS Number: 80222-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR