Glycinamide hydrochloride (H-Gly-NH2.HCl)

Supplier: Matrix Scientific

Synonyms: H-Gly-NH2.HCl

052747-25G 052747-100G
101900-876EA 21.54 USD
101900-876 101900-878
Glycinamide hydrochloride (H-Gly-NH2.HCl)
Glycinamide hydrochloride (H-Gly-NH2.HCl)

Hazard: Irritant/Hygroscopic

Formula: C₂H₇ClN₂O
MW: 110.54 g/mol
MDL Number: MFCD00013008
CAS Number: 1668-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR