3-Amino-4-chloro-N-ethylbenzenesulfonamide

Supplier: Matrix Scientific

052742-500MG 052742-2.5G
101900-854EA 364.62 USD
101900-854 101900-856
3-Amino-4-chloro-N-ethylbenzenesulfonamide
3-Amino-4-chloro-N-ethylbenzenesulfonamide

Hazard: Irritant

Formula: C₈H₁₁ClN₂O₂S
MDL Number: MFCD09900692
CAS Number: 1017477-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR