N-Ethyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052740-500MG 052740-2.5G
101900-846EA 166.15 USD
101900-846 101900-848
N-Ethyl-4-piperidinecarboxamide hydrochloride
N-Ethyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD13368255
CAS Number: 1981-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR