N-Ethyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052739-500MG 052739-2.5G
101900-842EA 364.62 USD
101900-842 101900-844
N-Ethyl-3-piperidinecarboxamide hydrochloride
N-Ethyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD13562352
CAS Number: 112989-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR