N-Ethyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052737-500MG 052737-2.5G
101900-834EA 364.62 USD
101900-834 101900-836
N-Ethyl-2-pyrrolidinecarboxamide hydrochloride
N-Ethyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MDL Number: MFCD13562350
CAS Number: 59179-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR