2-Amino-N-ethyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052736-500MG 052736-2.5G
101900-830EA 364.62 USD
101900-830 101900-832
2-Amino-N-ethyl-3-phenylpropanamide hydrochloride
2-Amino-N-ethyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O
MDL Number: MFCD13562349
CAS Number: 1246172-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR