2-Amino-N-ethyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052734-500MG 052734-2.5G
101900-822EA 166.15 USD
101900-822 101900-824
2-Amino-N-ethyl-3-methylbutanamide hydrochloride
2-Amino-N-ethyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13562347
CAS Number: 1236254-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR