2-Amino-N-ethylpropanamide hydrochloride
Supplier: Matrix Scientific
052732-500MG
052732-2.5G
101900-814EA
364.62
USD
101900-814
101900-816
2-Amino-N-ethylpropanamide hydrochloride
2-Amino-N-ethylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₅H₁₃ClN₂O |
MDL Number:
MFCD09971350 CAS Number: 1078162-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...