6-Chloro-N-ethyl-2-pyridinamine

Supplier: Matrix Scientific

052730-500MG 052730-2.5G
101900-806EA 364.62 USD
101900-806 101900-808
6-Chloro-N-ethyl-2-pyridinamine
6-Chloro-N-ethyl-2-pyridinamine

Hazard: Irritant

Formula: C₇H₉ClN₂
MDL Number: MFCD13562345
CAS Number: 89026-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR