5-Bromo-N-ethyl-2-pyridinamine

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-ethylaminopyridine

052729-500MG 052729-2.5G
101900-802EA 364.62 USD
101900-802 101900-804
5-Bromo-N-ethyl-2-pyridinamine
5-Bromo-N-ethyl-2-pyridinamine

Hazard: Irritant

Formula: C₇H₉BrN₂
MW: 201.07 g/mol
MDL Number: MFCD09743642
CAS Number: 856850-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR