N2-Ethyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052727-500MG 052727-2.5G
101900-794EA 364.62 USD
101900-794 101900-796
N2-Ethyl-2,3-pyridinediamine
N2-Ethyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₇H₁₁N₃
MDL Number: MFCD08692123
CAS Number: 32282-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR