2-(Ethylamino)isonicotinic acid
Supplier: Matrix Scientific
052725-500MG
052725-2.5G
101900-786EA
364.62
USD
101900-786
101900-788
2-(Ethylamino)isonicotinic acid
2-(Ethylamino)isonicotinic acid
Hazard: Irritant
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol |
MDL Number:
MFCD04115720 CAS Number: 86649-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...