2-Chloro-6-(ethylamino)pyrazine
Supplier: Matrix Scientific
052724-500MG
052724-2.5G
101900-782EA
364.62
USD
101900-782
101900-784
2-Chloro-6-(ethylamino)pyrazine
2-Chloro-6-(ethylamino)pyrazine
Hazard: Irritant
Formula:
C₈H₁₂ClN₃ |
MDL Number:
MFCD09878382 CAS Number: 957065-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...