6-Chloro-N-ethyl-4-pyrimidinamine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N-ethylpyrimidin-4-amine

052722-500MG 052722-2.5G
101900-774EA 364.62 USD
101900-774 101900-776
6-Chloro-N-ethyl-4-pyrimidinamine
6-Chloro-N-ethyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₆H₈ClN₃
MW: 157.6 g/mol
MDL Number: MFCD09475892
CAS Number: 872511-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR