N-(5-Bromo-4-methyl-2-pyridinyl)-N-ethylamine

Supplier: Matrix Scientific

052721-500MG 052721-2.5G
101900-770EA 364.62 USD
101900-770 101900-772
N-(5-Bromo-4-methyl-2-pyridinyl)-N-ethylamine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-ethylamine

Hazard: Irritant

Formula: C₈H₁₁BrN₂
MDL Number: MFCD13562343
CAS Number: 1220036-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR