4-(Ethylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

052720-500MG 052720-2.5G
101900-766EA 364.62 USD
101900-766 101900-768
4-(Ethylamino)-3-nitrobenzoic acid
4-(Ethylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₄
MDL Number: MFCD00546760
CAS Number: 2788-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR