Ethyl-3-amino-4-(ethylamino)benzoate

Supplier: Matrix Scientific

052719-500MG 052719-2.5G
101900-762EA 364.62 USD
101900-762 101900-764
Ethyl-3-amino-4-(ethylamino)benzoate
Ethyl-3-amino-4-(ethylamino)benzoate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD00187201
CAS Number: 202131-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR