3-Amino-4-chloro-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

052718-500MG 052718-2.5G
101900-758EA 364.62 USD
101900-758 101900-760
3-Amino-4-chloro-N-methylbenzenesulfonamide
3-Amino-4-chloro-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₇H₉ClN₂O₂S
MDL Number: MFCD09900665
CAS Number: 41607-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR