N-Methyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

Synonyms: N-Methylpiperidine-4-carboxamide hydrochloride

052716-500MG 052716-2.5G
101900-750EA 364.62 USD
101900-750 101900-752
N-Methyl-4-piperidinecarboxamide hydrochloride
N-Methyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD11615742
CAS Number: 1903-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR