N-Methyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

Synonyms: N-Methylpiperidine-3-carboxamide hydrochloride

052715-500MG 052715-2.5G
101900-746EA 364.62 USD
101900-746 101900-748
N-Methyl-3-piperidinecarboxamide hydrochloride
N-Methyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD09064923
CAS Number: 475060-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR