N-Methyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

Synonyms: N-Methyl-2-pyrrolidinecarboxamide HCl

052713-500MG 052713-2.5G
101900-738EA 364.62 USD
101900-738 101900-740
N-Methyl-2-pyrrolidinecarboxamide hydrochloride
N-Methyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₂N₂O·ClH
MW: 164.633 g/mol
MDL Number: MFCD11858066
CAS Number: 137693-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR