2-Amino-N-methyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052712-500MG 052712-2.5G
101900-734EA 147.69 USD
101900-734 101900-736
2-Amino-N-methyl-3-phenylpropanamide hydrochloride
2-Amino-N-methyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MW: 214.7 g/mol
MDL Number: MFCD13562340
CAS Number: 1236261-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR