2-Amino-N-3-dimethylbutanamide hydrochloride

Supplier: Matrix Scientific

052710-500MG 052710-2.5G
101900-726EA 384.62 USD
101900-726 101900-728
2-Amino-N-3-dimethylbutanamide hydrochloride
2-Amino-N-3-dimethylbutanamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O
MDL Number: MFCD13562338
CAS Number: 188890-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR