3-Amino-N-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052709-500MG 052709-2.5G
101900-722EA 323.08 USD
101900-722 101900-724
3-Amino-N-methylpropanamide hydrochloride
3-Amino-N-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O
MDL Number: MFCD10686841
CAS Number: 51739-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR