2-Amino-N-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052707-500MG 052707-2.5G
101900-714EA 40 USD
101900-714 101900-716
2-Amino-N-methylpropanamide hydrochloride
2-Amino-N-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O
MDL Number: MFCD13562337
CAS Number: 38215-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR