5-Bromo-N-methylpyridin-2-amine
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-(N-methylamino)pyridine, 5-Bromo-2-methylaminopyridine
052704-500MG
052704-2.5G
101900-702EA
50.77
USD
101900-702
101900-704
5-Bromo-N-methylpyridin-2-amine
5-Bromo-N-methylpyridin-2-amine
Hazard: Irritant
Formula:
C₆H₇BrN₂ MW: 187.0400 g/mol |
MDL Number:
MFCD08272096 CAS Number: 84539-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...