5-Bromo-N,3-dimethyl-2-pyridinamine

Supplier: Matrix Scientific

052703-500MG 052703-2.5G
101900-698EA 364.62 USD
101900-698 101900-700
5-Bromo-N,3-dimethyl-2-pyridinamine
5-Bromo-N,3-dimethyl-2-pyridinamine

Hazard: Irritant

Formula: C₇H₉BrN₂
MDL Number: MFCD10000482
CAS Number: 245765-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR