2-Chloro-N-methylnicotinamide

Supplier: Matrix Scientific

052702-500MG 052702-2.5G
101900-694EA 76.92 USD
101900-694 101900-696
2-Chloro-N-methylnicotinamide
2-Chloro-N-methylnicotinamide

Hazard: Irritant

Formula: C₇H₇ClN₂O
MDL Number: MFCD08691934
CAS Number: 52943-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR