2-Chloro-6-(methylamino)pyrazine
Supplier: Matrix Scientific
Synonyms:
6-Chloro-N-methylpyrazin-2-amine
052700-500MG
052700-2.5G
101900-686EA
364.62
USD
101900-686
101900-688
2-Chloro-6-(methylamino)pyrazine
2-Chloro-6-(methylamino)pyrazine
Hazard: Irritant
Formula:
C₅H₆ClN₃ MW: 143.5700 g/mol |
MDL Number:
MFCD09878383 CAS Number: 848366-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...