3-Chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

Synonyms: 3-Chloro-N-methyl-5-(trifluoromethyl)pyridin-2-amine

052699-500MG 052699-2.5G
101900-682EA 364.62 USD
101900-682 101900-684
3-Chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-methyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₇H₆ClF₃N₂
MW: 210.59 g/mol
MDL Number: MFCD06375956
CAS Number: 89810-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR