5-Bromo-2-(methylamino)pyridine

Supplier: Matrix Scientific

052697-500MG 052697-2.5G
101900-674EA 364.62 USD
101900-674 101900-676
5-Bromo-2-(methylamino)pyridine
5-Bromo-2-(methylamino)pyridine

Hazard: Irritant

Formula: C₇H₉BrN₂
MDL Number: MFCD07375098
CAS Number: 155789-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR