Ethyl-3-amino-4-(methylamino)benzoate
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-amino-4-(methylamino)benzoate
052695-500MG
052695-2.5G
101900-666EA
50.77
USD
101900-666
101900-668
Ethyl-3-amino-4-(methylamino)benzoate
Ethyl-3-amino-4-(methylamino)benzoate
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₂ MW: 194.24 g/mol |
MDL Number:
MFCD04110945 CAS Number: 66315-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...