2-{[Benzyl(ethyl)amino]methyl}-4-fluoroaniline

Supplier: Matrix Scientific

052693-500MG 052693-2.5G
101900-658EA 364.62 USD
101900-658 101900-660
2-{[Benzyl(ethyl)amino]methyl}-4-fluoroaniline
2-{[Benzyl(ethyl)amino]methyl}-4-fluoroaniline

Hazard: Irritant

Formula: C₁₆H₁₉FN₂
MDL Number: MFCD12577680
CAS Number: 1153197-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR