2-(2,3-Dihydro-1H-indol-1-ylmethyl)-4-fluorophenylamine

Supplier: Matrix Scientific

052691-500MG 052691-2.5G
101900-650EA 364.62 USD
101900-650 101900-652
2-(2,3-Dihydro-1H-indol-1-ylmethyl)-4-fluorophenylamine
2-(2,3-Dihydro-1H-indol-1-ylmethyl)-4-fluorophenylamine

Hazard: Irritant

Formula: C₁₅H₁₅FN₂
MDL Number: MFCD11980010
CAS Number: 1154634-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR