2-{[Cyclohexyl(ethyl)amino]methyl}-4-fluoroaniline

Supplier: Matrix Scientific

052688-500MG 052688-2.5G
101900-638EA 364.62 USD
101900-638 101900-640
2-{[Cyclohexyl(ethyl)amino]methyl}-4-fluoroaniline
2-{[Cyclohexyl(ethyl)amino]methyl}-4-fluoroaniline

Hazard: Irritant

Formula: C₁₅H₂₃FN₂
MDL Number: MFCD11980008
CAS Number: 1154634-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR