2-{[Cyclohexyl(methyl)amino]methyl}-4-fluoroaniline

Supplier: Matrix Scientific

052685-500MG 052685-2.5G
101900-626EA 364.62 USD
101900-626 101900-628
2-{[Cyclohexyl(methyl)amino]methyl}-4-fluoroaniline
2-{[Cyclohexyl(methyl)amino]methyl}-4-fluoroaniline

Hazard: Irritant

Formula: C₁₄H₂₁FN₂
MDL Number: MFCD12411439
CAS Number: 57814-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR