2-[(Dipropylamino)methyl]-4-fluoroaniline

Supplier: Matrix Scientific

052684-500MG 052684-2.5G
101900-622EA 364.62 USD
101900-622 101900-624
2-[(Dipropylamino)methyl]-4-fluoroaniline
2-[(Dipropylamino)methyl]-4-fluoroaniline

Hazard: Irritant

Formula: C₁₃H₂₁FN₂
MDL Number: MFCD12171385
CAS Number: 1156224-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR