1-(2-Amino-5-fluorobenzyl)-4-piperidinol

Supplier: Matrix Scientific

052683-500MG 052683-2.5G
101900-618EA 364.62 USD
101900-618 101900-620
1-(2-Amino-5-fluorobenzyl)-4-piperidinol
1-(2-Amino-5-fluorobenzyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₂H₁₇FN₂O
MDL Number: MFCD12171386
CAS Number: 1153396-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR