4-(2-Amino-5-fluorobenzyl)-2-piperazinone

Supplier: Matrix Scientific

052682-500MG 052682-2.5G
101900-614EA 364.62 USD
101900-614 101900-616
4-(2-Amino-5-fluorobenzyl)-2-piperazinone
4-(2-Amino-5-fluorobenzyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₁H₁₄FN₃O
MDL Number: MFCD12171384
CAS Number: 1153197-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR