4-Fluoro-2-[(4-methyl-1-piperidinyl)methyl]aniline

Supplier: Matrix Scientific

052678-500MG 052678-2.5G
101900-598EA 364.62 USD
101900-598 101900-600
4-Fluoro-2-[(4-methyl-1-piperidinyl)methyl]aniline
4-Fluoro-2-[(4-methyl-1-piperidinyl)methyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₉FN₂
MDL Number: MFCD12171371
CAS Number: 1153395-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR