4-Fluoro-2-[(3-methyl-1-piperidinyl)methyl]aniline

Supplier: Matrix Scientific

052677-500MG 052677-2.5G
101900-594EA 364.62 USD
101900-594 101900-596
4-Fluoro-2-[(3-methyl-1-piperidinyl)methyl]aniline
4-Fluoro-2-[(3-methyl-1-piperidinyl)methyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₉FN₂
MDL Number: MFCD12171378
CAS Number: 1153197-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR