4-Fluoro-2-[(2-methyl-1-piperidinyl)methyl]aniline

Supplier: Matrix Scientific

052676-500MG 052676-2.5G
101900-590EA 364.62 USD
101900-590 101900-592
4-Fluoro-2-[(2-methyl-1-piperidinyl)methyl]aniline
4-Fluoro-2-[(2-methyl-1-piperidinyl)methyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₉FN₂
MDL Number: MFCD12171375
CAS Number: 1153197-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR