2-[(2-Amino-5-fluorobenzyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

052674-500MG 052674-2.5G
101900-582EA 364.62 USD
101900-582 101900-584
2-[(2-Amino-5-fluorobenzyl)(ethyl)amino]-1-ethanol
2-[(2-Amino-5-fluorobenzyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₇FN₂O
MDL Number: MFCD12813609
CAS Number: 1182922-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR