2-{[Butyl(methyl)amino]methyl}-4-fluoroaniline

Supplier: Matrix Scientific

052673-500MG 052673-2.5G
101900-578EA 364.62 USD
101900-578 101900-580
2-{[Butyl(methyl)amino]methyl}-4-fluoroaniline
2-{[Butyl(methyl)amino]methyl}-4-fluoroaniline

Hazard: Irritant

Formula: C₁₂H₁₉FN₂
MDL Number: MFCD12171390
CAS Number: 1156225-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR