1-(2-Amino-5-fluorobenzyl)-3-pyrrolidinol
Supplier: Matrix Scientific
052672-500MG
052672-2.5G
101900-574EA
364.62
USD
101900-574
101900-576
1-(2-Amino-5-fluorobenzyl)-3-pyrrolidinol
1-(2-Amino-5-fluorobenzyl)-3-pyrrolidinol
Hazard: Irritant
Formula:
C₁₁H₁₅FN₂O |
MDL Number:
MFCD13562333 CAS Number: 1219957-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...