4-Fluoro-2-(1-piperidinylmethyl)aniline

Supplier: Matrix Scientific

052670-500MG 052670-2.5G
101900-566EA 364.62 USD
101900-566 101900-568
4-Fluoro-2-(1-piperidinylmethyl)aniline
4-Fluoro-2-(1-piperidinylmethyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MW: 208.28 g/mol
MDL Number: MFCD12171383
CAS Number: 1153197-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR