2-[(Diethylamino)methyl]-4-fluoroaniline
Supplier: Matrix Scientific
052668-500MG
052668-2.5G
101900-558EA
364.62
USD
101900-558
101900-560
2-[(Diethylamino)methyl]-4-fluoroaniline
2-[(Diethylamino)methyl]-4-fluoroaniline
Hazard: Irritant
Formula:
C₁₁H₁₇FN₂ |
MDL Number:
MFCD12171374 CAS Number: 1153395-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...