4-Fluoro-2-(1-pyrrolidinylmethyl)aniline

Supplier: Matrix Scientific

052667-500MG 052667-2.5G
101900-554EA 364.62 USD
101900-554 101900-556
4-Fluoro-2-(1-pyrrolidinylmethyl)aniline
4-Fluoro-2-(1-pyrrolidinylmethyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₅FN₂
MDL Number: MFCD12171372
CAS Number: 1153395-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR