3-Amino-4-chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)benzenesulfonamide

Supplier: Matrix Scientific

052665-500MG 052665-2.5G
101900-546EA 364.62 USD
101900-546 101900-548
3-Amino-4-chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)benzenesulfonamide
3-Amino-4-chloro-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃S
MDL Number: MFCD13562332
CAS Number: 1220027-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR